Anti-APH1A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:50-1:200
Type:
Antigen: APH1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-756
- 77725-755
- Antigen Symbol:
- APH1A
- APH1A
- Antigen Synonyms:
- APH-1a|Likely ortholog of C. elegans anterior pharynx defective 1A|CGI78|Anterior Pharynx Defective 1|Presenilin Stabilization Factor|Aph1a|Aph-1alpha|Anterior pharynx defective 1 homolog A|Gamma Secretase Subunit APH1a|6530402N02Rik|APH 1A|Gamma secretase subunit APH 1A|CGI 78|AL138795.3|PSF|Presenilin-stabilization factor|APH1A gamma secretase subunit|Gamma-secretase subunit APH-1A|UNQ579/PRO1141|Aph 1alpha|APH1A_HUMAN
- APH-1a|Likely ortholog of C. elegans anterior pharynx defective 1A|CGI78|Anterior Pharynx Defective 1|Presenilin Stabilization Factor|Aph1a|Aph-1alpha|Anterior pharynx defective 1 homolog A|Gamma Secretase Subunit APH1a|6530402N02Rik|APH 1A|Gamma secretase subunit APH 1A|CGI 78|AL138795.3|PSF|Presenilin-stabilization factor|APH1A gamma secretase subunit|Gamma-secretase subunit APH-1A|UNQ579/PRO1141|Aph 1alpha|APH1A_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96BI3
- Q96BI3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Gamma-secretase subunit APH-1A protein (90-118AA)
- Recombinant Human Gamma-secretase subunit APH-1A protein (90-118AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together