Anti-SLC35B4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF:1:50-1:200
Type:
Antigen: SLC35B4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-709
- 77725-708
- Antigen Symbol:
- SLC35B4
- SLC35B4
- Antigen Synonyms:
- YEA4|YEA4 homolog|UDP-xylose and UDP-N-acetylglucosamine transporter|Solute carrier family 35 member B4|SLC35B4|PSEC0055
- YEA4|YEA4 homolog|UDP-xylose and UDP-N-acetylglucosamine transporter|Solute carrier family 35 member B4|SLC35B4|PSEC0055
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969S0
- Q969S0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human UDP-xylose and UDP-N-acetylglucosamine transporter protein (174-200AA)
- Recombinant Human UDP-xylose and UDP-N-acetylglucosamine transporter protein (174-200AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together