Anti-MR1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: MR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-675
- 77725-674
- Antigen Symbol:
- MR1
- MR1
- Antigen Synonyms:
- MHC class I related gene protein|Class I histocompatibility antigen like protein|Major histocompatibility complex class I related isoform CRA d|Mr1|MHC class I related protein 1 isoform C|MHC class I like antigen MR 1|Major histocompatibility complex class I related gene protein|Mr1 protein|MHC class I like antigen MR1|MHC class I-related gene protein|MR1C|Major histocompatibility complex class I like sequence|Major histocompatibility complex class I-related gene protein|Major histocompatibility complex class I related isoform CRA a|MHC class I related protein 1 isoform B|Major histocompatibility complex class I related isoform CRA c|HMR1_HUMAN|MR 1|Major histocompatibility complex class I related|Class I histocompatibility antigen-like protein|HLALS|MR1B
- MHC class I related gene protein|Class I histocompatibility antigen like protein|Major histocompatibility complex class I related isoform CRA d|Mr1|MHC class I related protein 1 isoform C|MHC class I like antigen MR 1|Major histocompatibility complex class I related gene protein|Mr1 protein|MHC class I like antigen MR1|MHC class I-related gene protein|MR1C|Major histocompatibility complex class I like sequence|Major histocompatibility complex class I-related gene protein|Major histocompatibility complex class I related isoform CRA a|MHC class I related protein 1 isoform B|Major histocompatibility complex class I related isoform CRA c|HMR1_HUMAN|MR 1|Major histocompatibility complex class I related|Class I histocompatibility antigen-like protein|HLALS|MR1B
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q95460
- Q95460
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Major histocompatibility complex class I-related gene protein (20-260AA)
- Recombinant Human Major histocompatibility complex class I-related gene protein (20-260AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together