Anti-MED12 Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: MED12
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77725-668
- 77725-669
- Antigen Symbol:
- MED12
- MED12
- Antigen Synonyms:
- CAG repeat protein 45|Trinucleotide repeat-containing gene 11 protein|OPA containing protein|CAGH45|Activator recruited cofactor 240 kDa component|Mediator of RNA polymerase II transcription subunit 12|MED12|Trap230|ARC240|Activator-recruited cofactor 240 kDa component|Mediator complex subunit 12|MED12_HUMAN|KIAA0192|Trinucleotide repeat containing gene 11 protein|HOPA|Thyroid hormone receptor associated protein complex 230 kDa component|TNRC11|OPA-containing protein|Thyroid hormone receptor-associated protein complex 230 kDa component
- CAG repeat protein 45|Trinucleotide repeat-containing gene 11 protein|OPA containing protein|CAGH45|Activator recruited cofactor 240 kDa component|Mediator of RNA polymerase II transcription subunit 12|MED12|Trap230|ARC240|Activator-recruited cofactor 240 kDa component|Mediator complex subunit 12|MED12_HUMAN|KIAA0192|Trinucleotide repeat containing gene 11 protein|HOPA|Thyroid hormone receptor associated protein complex 230 kDa component|TNRC11|OPA-containing protein|Thyroid hormone receptor-associated protein complex 230 kDa component
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q93074
- Q93074
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Mediator of RNA polymerase II transcription subunit 12 protein (1742-2011AA)
- Recombinant Human Mediator of RNA polymerase II transcription subunit 12 protein (1742-2011AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together