Anti-EZH1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: EZH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-630
- 77725-631
- Antigen Symbol:
- EZH1
- EZH1
- Antigen Synonyms:
- Histone-lysine N-methyltransferase EZH1|EZH1_HUMAN|Enhancer of zeste homolog 1|Enhancer of zeste homolog 1 (Drosophila)|ENX-2|Ezh1|ENX2
- Histone-lysine N-methyltransferase EZH1|EZH1_HUMAN|Enhancer of zeste homolog 1|Enhancer of zeste homolog 1 (Drosophila)|ENX-2|Ezh1|ENX2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92800
- Q92800
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Histone-lysine N-methyltransferase EZH1 protein (160-280AA)
- Recombinant Human Histone-lysine N-methyltransferase EZH1 protein (160-280AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together