Anti-SNX33 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:500-1:1000
Type:
Antigen: SNX33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-483
- 77725-482
- Antigen Symbol:
- SNX33
- SNX33
- Antigen Synonyms:
- SH3 and PX domain containing protein 3|SNX33_HUMAN|SH3 and PX domain-containing protein 3|SNX30|SH3PX3|Sorting nexin 33|SH3PXD3C|SNX 33|SNX 30|Sorting nexin-33|snx33|SH3 and PX domain containing 3|MGC32065
- SH3 and PX domain containing protein 3|SNX33_HUMAN|SH3 and PX domain-containing protein 3|SNX30|SH3PX3|Sorting nexin 33|SH3PXD3C|SNX 33|SNX 30|Sorting nexin-33|snx33|SH3 and PX domain containing 3|MGC32065
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WV41
- Q8WV41
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sorting nexin-33 protein (348-516AA)
- Recombinant Human Sorting nexin-33 protein (348-516AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together