Anti-STK32A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:200-1:1000, IHC:1:20-1:200
Type:
Antigen: STK32A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77690-453
- 77725-445
- Antigen Symbol:
- STK32A
- STK32A
- Antigen Synonyms:
- YANK 1|ST32A_HUMAN|STK 32A|A930015B13Rik|MGC22688|Yet another novel kinase 1|Serine/threonine protein kinase 32A|Serine/threonine-protein kinase 32A|Serine/threonine kinase 32A|Stk32a|YANK1
- YANK 1|ST32A_HUMAN|STK 32A|A930015B13Rik|MGC22688|Yet another novel kinase 1|Serine/threonine protein kinase 32A|Serine/threonine-protein kinase 32A|Serine/threonine kinase 32A|Stk32a|YANK1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8WU08
- Q8WU08
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase 32A protein (1-166AA)
- Recombinant Human Serine/threonine-protein kinase 32A protein (1-166AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together