Anti-ARX Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARX
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77725-015
- 77725-014
- Antigen Symbol:
- ARX
- ARX
- Antigen Synonyms:
- Arx|ARX_HUMAN|MRXS1|Homeobox protein ARX|MRX54|Aristaless-related homeobox|ISSX|PRTS|phox2a|Aristaless related homeobox gene|MRX43|MRX32|MRX36|MRX38|MRX29|MRX33
- Arx|ARX_HUMAN|MRXS1|Homeobox protein ARX|MRX54|Aristaless-related homeobox|ISSX|PRTS|phox2a|Aristaless related homeobox gene|MRX43|MRX32|MRX36|MRX38|MRX29|MRX33
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96QS3
- Q96QS3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Homeobox protein ARX protein (381-460AA)
- Recombinant Human Homeobox protein ARX protein (381-460AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together