Anti-DGAT2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DGAT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-985
- 77724-986
- Antigen Symbol:
- DGAT2
- DGAT2
- Antigen Synonyms:
- Diacylglycerol O acyltransferase like protein 2|Diacylglycerol O-acyltransferase-like protein 2 isoform 1|Diacylglycerol O-acyltransferase homolog 2|GS1999full|HMFN1045|Diglyceride acyltransferase 2|DGAT2|DGAT2_HUMAN|DKFZp686A15125|Diacylglycerol O-acyltransferase 2|Diacylglycerol O-acyltransferase homolog 2 (mouse)
- Diacylglycerol O acyltransferase like protein 2|Diacylglycerol O-acyltransferase-like protein 2 isoform 1|Diacylglycerol O-acyltransferase homolog 2|GS1999full|HMFN1045|Diglyceride acyltransferase 2|DGAT2|DGAT2_HUMAN|DKFZp686A15125|Diacylglycerol O-acyltransferase 2|Diacylglycerol O-acyltransferase homolog 2 (mouse)
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96PD7
- Q96PD7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Diacylglycerol O-acyltransferase 2 protein (1-66AA)
- Recombinant Human Diacylglycerol O-acyltransferase 2 protein (1-66AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together