Anti-WDFY2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IF:1:50-1:200
Type:
Antigen: WDFY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-978
- 77690-421
- Antigen Symbol:
- WDFY2
- WDFY2
- Antigen Synonyms:
- WD repeat and FYVE domain-containing protein 2|ZFYVE22|WD40- and FYVE domain-containing protein 2|WDFY2_HUMAN|Wdfy2|WD40 and FYVE domain containing 2|WD repeat and FYVE domain containing 2|WDF2|Zinc finger FYVE domain-containing protein 22|Propeller-FYVE protein|PROF|RP11 147H23.1
- WD repeat and FYVE domain-containing protein 2|ZFYVE22|WD40- and FYVE domain-containing protein 2|WDFY2_HUMAN|Wdfy2|WD40 and FYVE domain containing 2|WD repeat and FYVE domain containing 2|WDF2|Zinc finger FYVE domain-containing protein 22|Propeller-FYVE protein|PROF|RP11 147H23.1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96P53
- Q96P53
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human WD repeat and FYVE domain-containing protein 2 protein (2-114AA)
- Recombinant Human WD repeat and FYVE domain-containing protein 2 protein (2-114AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together