Anti-CDCA7L Rabbit Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: CDCA7L
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77724-849
- 77724-850
- Antigen Symbol:
- CDCA7L
- CDCA7L
- Antigen Synonyms:
- Cdca7l|Cell division cycle-associated 7-like protein|CDA7L_HUMAN|RAM2|HR1|Transcription factor RAM2|c Myc Interacting protein JPO2|Protein JPO2|DKFZp762L0311|R1|JPO2
- Cdca7l|Cell division cycle-associated 7-like protein|CDA7L_HUMAN|RAM2|HR1|Transcription factor RAM2|c Myc Interacting protein JPO2|Protein JPO2|DKFZp762L0311|R1|JPO2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96GN5
- Q96GN5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Cell division cycle-associated 7-like protein (14-167AA)
- Recombinant Human Cell division cycle-associated 7-like protein (14-167AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together