Anti-PIK3IP1 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIK3IP1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-814
- 77724-813
- Antigen Symbol:
- PIK3IP1
- PIK3IP1
- Antigen Synonyms:
- Kringle domain containing protein HGFL|hHGFL(S)|Kringle domain-containing protein HGFL|PIK3IP1|HGFL|MGC17330|Phosphoinositide 3 kinase interacting protein 1|Phosphoinositide-3-kinase-interacting protein 1|P3IP1_HUMAN
- Kringle domain containing protein HGFL|hHGFL(S)|Kringle domain-containing protein HGFL|PIK3IP1|HGFL|MGC17330|Phosphoinositide 3 kinase interacting protein 1|Phosphoinositide-3-kinase-interacting protein 1|P3IP1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96FE7
- Q96FE7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Phosphoinositide-3-kinase-interacting protein 1 protein (22-168AA)
- Recombinant Human Phosphoinositide-3-kinase-interacting protein 1 protein (22-168AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together