Anti-NSUN4 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: NSUN4
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77724-759
- 77724-760
- Antigen Symbol:
- NSUN4
- NSUN4
- Antigen Synonyms:
- EC=2.1.1.-|NSUN4_HUMAN|Nsun4|NOP2/Sun domain family, member 4|5 methylcytosine rRNA methyltransferase NSUN4|Shtap|NOL1/NOP2/Sun domain family member 4|NOL1/NOP2/Sun domain family, member 4|Sperm head and tail associated protein|Putative methyltransferase NSUN4|RP4-603I14.2
- EC=2.1.1.-|NSUN4_HUMAN|Nsun4|NOP2/Sun domain family, member 4|5 methylcytosine rRNA methyltransferase NSUN4|Shtap|NOL1/NOP2/Sun domain family member 4|NOL1/NOP2/Sun domain family, member 4|Sperm head and tail associated protein|Putative methyltransferase NSUN4|RP4-603I14.2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96CB9
- Q96CB9
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human 5-methylcytosine rRNA methyltransferase NSUN4 protein (26-284AA)
- Recombinant Human 5-methylcytosine rRNA methyltransferase NSUN4 protein (26-284AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together