Anti-PEX13 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PEX13
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77724-622
- 77724-621
- Antigen Symbol:
- PEX13
- PEX13
- Antigen Synonyms:
- Peroxin-13|Peroxin 13|2610008O20Rik|Peroxisomal membrane protein PEX13|PEX 13|pex13|ZWS|Peroxisomal membrane protein PEX 13|Peroxisome biogenesis factor 13|NALD|PEX13_HUMAN|OTTMUSP00000005454|Peroxin13|RP23-188K3.1
- Peroxin-13|Peroxin 13|2610008O20Rik|Peroxisomal membrane protein PEX13|PEX 13|pex13|ZWS|Peroxisomal membrane protein PEX 13|Peroxisome biogenesis factor 13|NALD|PEX13_HUMAN|OTTMUSP00000005454|Peroxin13|RP23-188K3.1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q92968
- Q92968
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Peroxisomal membrane protein PEX13 protein (1-130AA)
- Recombinant Human Peroxisomal membrane protein PEX13 protein (1-130AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together