Anti-PIGX Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: PIGX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77724-375
- 77690-392
- Antigen Symbol:
- PIGX
- PIGX
- Antigen Synonyms:
- PIG-X|Phosphatidylinositol-glycan biosynthesis class X protein|Pigx|GPI mannosyltransferase subunit|PIGX_HUMAN|Phosphatidylinositol glycan class X
- PIG-X|Phosphatidylinositol-glycan biosynthesis class X protein|Pigx|GPI mannosyltransferase subunit|PIGX_HUMAN|Phosphatidylinositol glycan class X
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TBF5
- Q8TBF5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Phosphatidylinositol-glycan biosynthesis class X protein (42-230AA)
- Recombinant Human Phosphatidylinositol-glycan biosynthesis class X protein (42-230AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together