Anti-VANGL1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000, IHC:1:20-1:200
Type:
Antigen: VANGL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77690-390
- 77724-343
- Antigen Symbol:
- VANGL1
- VANGL1
- Antigen Synonyms:
- KAI1 C-terminal interacting tetraspanin|Drosophila|Vang-like 1|Vang like protein 1|VANG1_HUMAN|ANGL planar cell polarity protein 1|Strabismus|LPP2|STB2|VANGL 1|Vang like 1 (van gogh|Vang-like protein 1|Loop-tail protein 2 homolog|Loop tail protein 2 homolog|Van Gogh like protein 1|Strabismus 2|Vangl1|Drosophila)|1|2|homolog of|KITENIN|MGC5338|STBM2|Van Gogh|Van Gogh-like protein 1
- KAI1 C-terminal interacting tetraspanin|Drosophila|Vang-like 1|Vang like protein 1|VANG1_HUMAN|ANGL planar cell polarity protein 1|Strabismus|LPP2|STB2|VANGL 1|Vang like 1 (van gogh|Vang-like protein 1|Loop-tail protein 2 homolog|Loop tail protein 2 homolog|Van Gogh like protein 1|Strabismus 2|Vangl1|Drosophila)|1|2|homolog of|KITENIN|MGC5338|STBM2|Van Gogh|Van Gogh-like protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TAA9
- Q8TAA9
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Vang-like protein 1 protein (275-524AA)
- Recombinant Human Vang-like protein 1 protein (275-524AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together