Anti-ACSS1 (acetyl-K642) Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
WB:1:500-2000, ELISA:1:10000-20000
Type:
Antigen: ACSS1 (acetyl-K642)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77705-641
- 77724-342
- Antigen Symbol:
- ACSS1 (acetyl-K642)
- ACSS1 (acetyl-K642)
- Antigen Synonyms:
- ACAS2L|Acss1|Acetyl CoA synthetase 2|Acetyl Coenzyme A synthetase 2 (AMP forming) like|AceCS2L|FLJ45659|AceCS2|KIAA1846|Acetate--CoA ligase 2|Acyl-CoA synthetase short-chain family member 1|Acetyl-coenzyme A synthetase 2-like|mitochondrial|ACS2L_HUMAN|Acyl CoA synthetase short chain family member 1|Acetate CoA ligase 2|Acetyl coenzyme A synthetase 2 like|Acetyl-CoA synthetase 2|MGC33843
- ACAS2L|Acss1|Acetyl CoA synthetase 2|Acetyl Coenzyme A synthetase 2 (AMP forming) like|AceCS2L|FLJ45659|AceCS2|KIAA1846|Acetate--CoA ligase 2|Acyl-CoA synthetase short-chain family member 1|Acetyl-coenzyme A synthetase 2-like|mitochondrial|ACS2L_HUMAN|Acyl CoA synthetase short chain family member 1|Acetate CoA ligase 2|Acetyl coenzyme A synthetase 2 like|Acetyl-CoA synthetase 2|MGC33843
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NUB1
- Q9NUB1
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthesized Acetyl peptide derived from Human ACSS1. at AA range: K642
- Synthesized Acetyl peptide derived from Human ACSS1. at AA range: K642
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together