Anti-SLC22A18AS Rabbit Polyclonal Antibody (HRP)
New Product
Type:
Antigen: SLC22A18AS
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77724-083
- 77724-082
- Antigen Symbol:
- SLC22A18AS
- SLC22A18AS
- Antigen Synonyms:
- Organic cation transporter-like protein 2 antisense protein|BWR1B|p27-BWR1B|BWSCR1B|SLC22A1LSBeckwith-Wiedemann syndrome chromosomal region 1 candidate gene B protein|p27-Beckwith-Wiedemann region 1 B|SLC22A18AS|Solute carrier family 22 member 18 antisense protein|ORCTL2S|Solute carrier family 22 member 1-like antisense protein
- Organic cation transporter-like protein 2 antisense protein|BWR1B|p27-BWR1B|BWSCR1B|SLC22A1LSBeckwith-Wiedemann syndrome chromosomal region 1 candidate gene B protein|p27-Beckwith-Wiedemann region 1 B|SLC22A18AS|Solute carrier family 22 member 18 antisense protein|ORCTL2S|Solute carrier family 22 member 1-like antisense protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N1D0
- Q8N1D0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene B protein (1-150AA)
- Recombinant Human Beckwith-Wiedemann syndrome chromosomal region 1 candidate gene B protein (1-150AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together