Anti-mCherry-Tag Rabbit Polyclonal Antibody
77698-356
New Product
- Form:Liquid
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Protein
- Purification:Affinity purified
- Cat. No.:77698-356
- Packaging:Vial
WB:1:5000
Type:
Antigen: mCherry-Tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Multiple