Anti-Cleaved-ITIH2 (D702) Rabbit Polyclonal Antibody
Catalog # 77698-279
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Cleaved-ITIH2 (D702)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P19823
- Antigen Synonyms:Inter alpha trypsin inhibitor complex component II|AI747202|Serum derived hyaluronan associated protein|SHAP|Itih 2|ITIH2_HUMAN|Inter alpha trypsin inhibitor heavy chain H2|ITI heavy chain H2|IGHEP2|OTTHUMP00000019058|Inter-alpha-trypsin inhibitor complex component II|Serum-derived hyaluronan-associated protein|Inter alpha (globulin) inhibitor H2|Inter-alpha-trypsin inhibitor heavy chain H2|H2P|RP23 119N4.3|Itih2|ITI HC2|ITI-HC2|OTTHUMP00000019059|Inter-alpha-inhibitor heavy chain 2|Intin2|Inter alpha globulin inhibitor H2 polypeptide|OTTMUSP00000012019
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human ITI-H2
- Purification:Antigen affinity purified
- Cat. No.:77698-279
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:10000
Type:
Antigen: Cleaved-ITIH2 (D702)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human