Anti-Cleaved-ITGA5 (E895) Rabbit Polyclonal Antibody
Catalog # 77698-275
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Cleaved-ITGA5 (E895)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P08648
- Antigen Synonyms:FNRA|CD49e|Integrin alpha-5|Fibronectin receptor|Very late activation protein 5|alpha polypeptide)|CD49 antigen-like family member E|alpha subunit|Integrin alpha 5 (fibronectin receptor alpha)|ITA5_HUMAN|Integrin alpha-5 light chain|alpha 5 (fibronectin receptor|VLA-5|Integrin|Integrin alpha-F|VLA5|VLA5A|Fibronectin receptor subunit alpha|Itga5
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Integrin α5 LC
- Purification:Antigen affinity purified
- Cat. No.:77698-275
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: Cleaved-ITGA5 (E895)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human