Anti-Cleaved-MMP23A (Y79) Rabbit Polyclonal Antibody
77698-259
New Product
- Antigen Symbol:Cleaved-MMP23A (Y79)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O75900
- Antigen Synonyms:matrix metallopeptidase 23B|MMP23_HUMAN|MMP-23|MIFR|MMP 22|MMP-21|MMP-22|soluble form|Femalysin|matrix metalloproteinase 22|MIFR 1|MMP23B|MMP23|Matrix metalloproteinase-22|MIFR-1|Matrix metalloproteinase-23|matrix metalloproteinase 22 |matrix metalloproteinase in the female reproductive tract |MMP 21|Matrix metalloproteinase-21|Matrix metalloproteinase 23B|Matrix metalloproteinase 23|MMP23A |MMP21|MMP22|Matrix metallopeptidase 21|MMP 23|formerly
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human MMP-23
- Purification:Antigen affinity purified
- Cat. No.:77698-259
- Packaging:Vial
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: Cleaved-MMP23A (Y79)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat