Anti-Cleaved-BAD (D71) Rabbit Polyclonal Antibody
Catalog # 77698-242
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:Cleaved-BAD (D71)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q92934
- Antigen Synonyms:BBC 2|AI325008|Bcl 2 Binding Component 6|Bcl-2-like protein 8|Proapoptotic BH3 Only Protein|BCL2L8|BCL2 associated agonist of cell death|Bcl-2-binding component 6|Bcl2 Associated Death Promoter|BAD_HUMAN|Bcl2-L-8|BCL2 antagonist of cell death protein|BBC6|Bcl2 antagonist of cell death|BCL X / BCL 2 Binding Protein|BBC2|Bcl XL/Bcl 2 Associated Death Promoter|Bcl-XL/Bcl-2-associated death promoter|BCL2 binding protein|BCL X Binding Protein|BCL2 binding component 6|Bcl2 Like 8 Protein|BAD|Bcl 2 Antagonist of Cell Death
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Mouse Bad
- Purification:Antigen affinity purified
- Cat. No.:77698-242
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: Cleaved-BAD (D71)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat