Anti-Histone H3 (Di-methyl-K27) Mouse Monoclonal Antibody
77698-021
New Product
- Form:liquid
- Gene ID:P68431, Q71DI3, P84243
- Antigen Synonyms:H3FA|HIST1H3J|H3FJ|HIST1H3E|HIST1H3G|HIST1H3B|Histone H3/f|H3FI|HIST1H3D|HIST1H3C|HIST1H3I|HIST1H3F|Histone H3/b|Histone H3/d|Histone H3.1|H3FH|H3FD|Histone H3/a|H3FF|Histone H3/|HIST1H3H|H3FC|H3FB|H3FL|Histone H3/c|H3FK|HIST1H3A
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthetic Peptide
- Purification:Antigen affinity purified
- Cat. No.:77698-021
- Packaging:Vial
Recommended dilution: WB:1:500-1:5000
Type:
Antigen: Histone H3 (Di-methyl-K27)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat