Anti-Myc-Tag Mouse Monoclonal Antibody
Catalog # 77697-952
Supplier: AFG Bioscience
New Product
Specifications
- Form:Liquid
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthetic Peptide
- Purification:Affinity purified
- Cat. No.:77697-952
- Packaging:Vial
Specifications
About this item
Recommended dilution:WB:1:500-1:5000
Type:
Antigen: Myc-Tag
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Multiple