Anti-CD300LF Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CD300LF
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-664
- 77718-665
- Antigen Symbol:
- CD300LF
- CD300LF
- Antigen Synonyms:
- CD300LF|CMRF35-like molecule 1|IREM|IREM1|CLM-1|immune receptor expressed on myeloid cells|UNQ3105/PRO10111|Immune receptor expressed on myeloid cells 1|Inhibitory receptor IREM1|immunoglobin superfamily member|NKIR|Nepmucin|NK inhibitory receptor|IgSF13|CLM|Immunoglobulin superfamily member|CD300 antigen like family member|IGSF|CD300 antigen-like family member F|Immunoglobulin superfamily member 13|CLM1|IREM-1|CLM1_HUMAN|CD300 molecule like family member f|TREM|triggering receptors expressed on myeloid cells|CD300f
- CD300LF|CMRF35-like molecule 1|IREM|IREM1|CLM-1|immune receptor expressed on myeloid cells|UNQ3105/PRO10111|Immune receptor expressed on myeloid cells 1|Inhibitory receptor IREM1|immunoglobin superfamily member|NKIR|Nepmucin|NK inhibitory receptor|IgSF13|CLM|Immunoglobulin superfamily member|CD300 antigen like family member|IGSF|CD300 antigen-like family member F|Immunoglobulin superfamily member 13|CLM1|IREM-1|CLM1_HUMAN|CD300 molecule like family member f|TREM|triggering receptors expressed on myeloid cells|CD300f
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TDQ1
- Q8TDQ1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human CMRF35-like molecule 1 protein (20-156AA)
- Recombinant Human CMRF35-like molecule 1 protein (20-156AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together