Anti-MRPL30 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MRPL30
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-623
- 77718-622
- Antigen Symbol:
- MRPL30
- MRPL30
- Antigen Synonyms:
- RPML28|Mitochondrial large ribosomal subunit protein uL30m|L28mt|L30mt|HSPC249|39S ribosomal protein L30|MRPL30|MRPL28|mitochondrial|39S ribosomal protein L28|MRP-L28|MRP-L30
- RPML28|Mitochondrial large ribosomal subunit protein uL30m|L28mt|L30mt|HSPC249|39S ribosomal protein L30|MRPL30|MRPL28|mitochondrial|39S ribosomal protein L28|MRP-L28|MRP-L30
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TCC3
- Q8TCC3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human 39S ribosomal protein L30, mitochondrial protein (35-161AA)
- Recombinant Human 39S ribosomal protein L30, mitochondrial protein (35-161AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together