Anti-PIP4K2C Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIP4K2C
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-609
- 77718-608
- Antigen Symbol:
- PIP4K2C
- PIP4K2C
- Antigen Synonyms:
- Phosphatidylinositol-5-phosphate 4-kinase type-2 gamma|Phosphatidylinositol-5-phosphate 4-kinase type II gamma|PI42C_HUMAN|Phosphatidylinositol 4 phosphate 5 kinase|PIP4KII-gamma|Pip4k2c|PIP5K2C|PI(5)P 4-kinase type II gamma|Phosphatidylinositol 4 phosphate 5 kinase type II gamma
- Phosphatidylinositol-5-phosphate 4-kinase type-2 gamma|Phosphatidylinositol-5-phosphate 4-kinase type II gamma|PI42C_HUMAN|Phosphatidylinositol 4 phosphate 5 kinase|PIP4KII-gamma|Pip4k2c|PIP5K2C|PI(5)P 4-kinase type II gamma|Phosphatidylinositol 4 phosphate 5 kinase type II gamma
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TBX8
- Q8TBX8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Phosphatidylinositol 5-phosphate 4-kinase type-2 gamma protein (127-408AA )
- Recombinant Human Phosphatidylinositol 5-phosphate 4-kinase type-2 gamma protein (127-408AA )
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together