Anti-HSCB Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:20-1:200
Type:
Antigen: HSCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Specifications
- Catalog No:
- 77718-043
- 77690-089
- Antigen Symbol:
- HSCB
- HSCB
- Antigen Synonyms:
- DNAJC20|HSC20_HUMAN|dJ366L4.2|JAC1|Hsc20|AI325508|Iron-sulfur cluster co-chaperone protein HscB|AW049829|mitochondrial|hscB|J type co chaperone HSC20|RGD1311005|DnaJ homolog subfamily C member 20|RP3-366L4.2|Iron sulfur cluster co chaperone protein HscB mitochondrial|DnaJ (Hsp40) homolog, subfamily C member 20|HscB iron sulfur cluster co chaperone homolog
- DNAJC20|HSC20_HUMAN|dJ366L4.2|JAC1|Hsc20|AI325508|Iron-sulfur cluster co-chaperone protein HscB|AW049829|mitochondrial|hscB|J type co chaperone HSC20|RGD1311005|DnaJ homolog subfamily C member 20|RP3-366L4.2|Iron sulfur cluster co chaperone protein HscB mitochondrial|DnaJ (Hsp40) homolog, subfamily C member 20|HscB iron sulfur cluster co chaperone homolog
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IWL3
- Q8IWL3
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Iron-sulfur cluster co-chaperone protein HscB, mitochondrial protein (30-235AA)
- Recombinant Human Iron-sulfur cluster co-chaperone protein HscB, mitochondrial protein (30-235AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together