Anti-WDFY1 Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: WDFY1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77718-029
- 77718-030
- Antigen Symbol:
- WDFY1
- WDFY1
- Antigen Synonyms:
- WDF1|Phosphoinositide binding protein SR1|Phosphoinositide binding protein 1|Zinc finger FYVE domain containing protein 17|WD repeat and FYVE domain containing 1|FENS1|ZFYVE17|FENS-1|Phosphoinositide-binding protein 1|WD40 and FYVE domain containing protein 1|WD repeat and FYVE domain-containing protein 1|WDFY1|KIAA1435|FENS 1|Zinc finger FYVE domain-containing protein 17|WD40- and FYVE domain-containing protein 1|WDFY1_HUMAN
- WDF1|Phosphoinositide binding protein SR1|Phosphoinositide binding protein 1|Zinc finger FYVE domain containing protein 17|WD repeat and FYVE domain containing 1|FENS1|ZFYVE17|FENS-1|Phosphoinositide-binding protein 1|WD40 and FYVE domain containing protein 1|WD repeat and FYVE domain-containing protein 1|WDFY1|KIAA1435|FENS 1|Zinc finger FYVE domain-containing protein 17|WD40- and FYVE domain-containing protein 1|WDFY1_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IWB7
- Q8IWB7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human WD repeat and FYVE domain-containing protein 1 protein (261-357AA)
- Recombinant Human WD repeat and FYVE domain-containing protein 1 protein (261-357AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together