Anti-AEBP1 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: AEBP1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77717-757
- 77717-758
- Antigen Symbol:
- AEBP1
- AEBP1
- Antigen Synonyms:
- Adipocyte enhancer-binding protein 1|AE-binding protein 1|AEBP1|ACLP|Aortic carboxypeptidase-like protein
- Adipocyte enhancer-binding protein 1|AE-binding protein 1|AEBP1|ACLP|Aortic carboxypeptidase-like protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IUX7
- Q8IUX7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Adipocyte enhancer-binding protein 1 protein (46-234AA)
- Recombinant Human Adipocyte enhancer-binding protein 1 protein (46-234AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together