Anti-KRTAP8-1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: KRTAP8-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77690-067
- 77717-725
- Antigen Symbol:
- KRTAP8-1
- KRTAP8-1
- Antigen Synonyms:
- KRTAP8.1Keratin-associated protein 8-1|KRTAP8-1|High glycine-tyrosine keratin-associated protein 8.1|KAP8.1
- KRTAP8.1Keratin-associated protein 8-1|KRTAP8-1|High glycine-tyrosine keratin-associated protein 8.1|KAP8.1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IUC2
- Q8IUC2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Keratin-associated protein 8-1 protein (1-63AA)
- Recombinant Human Keratin-associated protein 8-1 protein (1-63AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together