Order Entry
United States
ContactUsLinkComponent
 
Anti-CCDC103 Rabbit Polyclonal Antibody (FITC)
New Product
 
undefined
Anti-CCDC103 Rabbit Polyclonal Antibody (FITC)
New Product

 

  • Catalog No:
  • 77717-610
  • 77717-609
  • Antigen Symbol:
  • CCDC103
  • CCDC103
  • Antigen Synonyms:
  • CCDC103Coiled-coil domain-containing protein 103
  • CCDC103Coiled-coil domain-containing protein 103
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q8IW40
  • Q8IW40
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Coiled-coil domain-containing protein 103 protein (1-242AA)
  • Recombinant Human Coiled-coil domain-containing protein 103 protein (1-242AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

 

Frequently Bought Together