Anti-LAX1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:500-1:5000, IF:1:50-1:200
Type:
Antigen: LAX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77690-057
- 77717-467
- Antigen Symbol:
- LAX1
- LAX1
- Antigen Synonyms:
- Linker for activation of X cells|LAX|LAX1|Membrane-associated adapter protein LAX|Lymphocyte transmembrane adapter 1
- Linker for activation of X cells|LAX|LAX1|Membrane-associated adapter protein LAX|Lymphocyte transmembrane adapter 1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IWV1
- Q8IWV1
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lymphocyte transmembrane adapter 1 protein (107-298AA)
- Recombinant Human Lymphocyte transmembrane adapter 1 protein (107-298AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together