Anti-GLB1L2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20-1:200
Type:
Antigen: GLB1L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77717-459
- 77717-460
- Antigen Symbol:
- GLB1L2
- GLB1L2
- Antigen Synonyms:
- GLBL2_HUMAN|UNQ210/PRO236|MST114|Galactosidase beta 1 like 2|MSTP014|Glb1l2|MSTP114|Beta-galactosidase-1-like protein 2
- GLBL2_HUMAN|UNQ210/PRO236|MST114|Galactosidase beta 1 like 2|MSTP014|Glb1l2|MSTP114|Beta-galactosidase-1-like protein 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IW92
- Q8IW92
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Beta-galactosidase-1-like protein 2 protein (401-636AA)
- Recombinant Human Beta-galactosidase-1-like protein 2 protein (401-636AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together