Anti-NIM1K Rabbit Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NIM1K
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77717-368
- 77717-367
- Antigen Symbol:
- NIM1K
- NIM1K
- Antigen Synonyms:
- NIM1|EC 2.7.11.1|NIM1 serine/threonine-protein kinase|Serine/threonine-protein kinase NIM1|NIM1K
- NIM1|EC 2.7.11.1|NIM1 serine/threonine-protein kinase|Serine/threonine-protein kinase NIM1|NIM1K
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IY84
- Q8IY84
- Isotype:
- IgG
- IgG
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase NIM1 protein (1-436AA)
- Recombinant Human Serine/threonine-protein kinase NIM1 protein (1-436AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together