Anti-SMC5 Rabbit Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SMC5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77717-353
- 77717-352
- Antigen Symbol:
- SMC5
- SMC5
- Antigen Synonyms:
- hSMC5|KIAA0594|Structural maintenance of chromosomes protein 5|SMC5L1|SMC protein 5|SMC-5|SMC5
- hSMC5|KIAA0594|Structural maintenance of chromosomes protein 5|SMC5L1|SMC protein 5|SMC-5|SMC5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IY18
- Q8IY18
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Structural maintenance of chromosomes protein 5 protein (296-403AA)
- Recombinant Human Structural maintenance of chromosomes protein 5 protein (296-403AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together