Anti-SCYL3 Rabbit Polyclonal Antibody (FITC)
New Product
Type:
Antigen: SCYL3
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77717-211
- 77717-210
- Antigen Symbol:
- SCYL3
- SCYL3
- Antigen Synonyms:
- PACE1 |SCY1 like 3|SCY1-like protein 3|SCY1 like protein 3|SCY1 like 3 (S. cerevisiae)|Protein-associating with the carboxyl-terminal domain of ezrin|SCY1 like pseudokinase 3|PACE 1 |SCYL 3|Ezrin binding protein PACE 1 |PACE1_HUMAN|Protein associating with the carboxyl terminal domain of ezrin|Ezrin-binding protein PACE-1|SCYL3
- PACE1 |SCY1 like 3|SCY1-like protein 3|SCY1 like protein 3|SCY1 like 3 (S. cerevisiae)|Protein-associating with the carboxyl-terminal domain of ezrin|SCY1 like pseudokinase 3|PACE 1 |SCYL 3|Ezrin binding protein PACE 1 |PACE1_HUMAN|Protein associating with the carboxyl terminal domain of ezrin|Ezrin-binding protein PACE-1|SCYL3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IZE3
- Q8IZE3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protein-associating with the carboxyl-terminal domain of ezrin protein (1-688AA)
- Recombinant Human Protein-associating with the carboxyl-terminal domain of ezrin protein (1-688AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together