Anti-TRIM59 Rabbit Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRIM59
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77717-110
- 77717-111
- Antigen Symbol:
- TRIM59
- TRIM59
- Antigen Synonyms:
- RING finger protein 104|MGC26631|Tripartite motif-containing protein 59|2700022F13Rik|Tumor suppressor TSBF-1|RNF104|tripartite motif containing 59|MGC129860|Tripartite motif containing 57|MGC129861|Tumor suppressor TSBF 1|TSBF1|TRIM57|TRIM59|TRI59_HUMAN|2310035M22Rik|MRF1
- RING finger protein 104|MGC26631|Tripartite motif-containing protein 59|2700022F13Rik|Tumor suppressor TSBF-1|RNF104|tripartite motif containing 59|MGC129860|Tripartite motif containing 57|MGC129861|Tumor suppressor TSBF 1|TSBF1|TRIM57|TRIM59|TRI59_HUMAN|2310035M22Rik|MRF1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8IWR1
- Q8IWR1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Tripartite motif-containing protein 59 protein (126-181AA)
- Recombinant Human Tripartite motif-containing protein 59 protein (126-181AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together