Anti-CXXC5 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
ELISA:1:2000-1:5000, IHC:1:50-1:200
Type:
Antigen: CXXC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77717-023
- 77717-022
- Antigen Symbol:
- CXXC5
- CXXC5
- Antigen Synonyms:
- CXXC-type zinc finger protein 5|CXXC finger protein 5|HSPC195|Putative MAPK-activating protein PM08|RINF|CF5|WT1-induced Inhibitor of Dishevelled|TCCCIA00297|CXXC5|retinoid-inducible nuclear factor|Putative NF-kappa-B-activating protein 102|CXXC5_HUMAN|CXXC finger 5 protein|WID
- CXXC-type zinc finger protein 5|CXXC finger protein 5|HSPC195|Putative MAPK-activating protein PM08|RINF|CF5|WT1-induced Inhibitor of Dishevelled|TCCCIA00297|CXXC5|retinoid-inducible nuclear factor|Putative NF-kappa-B-activating protein 102|CXXC5_HUMAN|CXXC finger 5 protein|WID
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7LFL8
- Q7LFL8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human CXXC5
- Fusion protein of Human CXXC5
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together