Anti-Wdpcp Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000-1:5000
Type:
Antigen: Wdpcp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 77690-031
- 77716-961
- Antigen Symbol:
- Wdpcp
- Wdpcp
- Antigen Synonyms:
- WdpcpWD repeat-containing and planar cell polarity effector protein fritz homolog|mFrtz|WD repeat-containing and planar cell polarity effector protein|Homolog-13
- WdpcpWD repeat-containing and planar cell polarity effector protein fritz homolog|mFrtz|WD repeat-containing and planar cell polarity effector protein|Homolog-13
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8C456
- Q8C456
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Mouse WD repeat-containing and planar cell polarity effector protein fritz homolog protein (1-722AA)
- Recombinant Mouse WD repeat-containing and planar cell polarity effector protein fritz homolog protein (1-722AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together