Order Entry
United States
ContactUsLinkComponent
Anti-GZMM Rabbit Polyclonal Antibody
Anti-GZMM Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-GZMM Rabbit Polyclonal Antibody
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77716-918
  • 77705-621
  • Antigen Symbol:
  • GZMM
  • GZMM
  • Antigen Synonyms:
  • GranzymeM|Gzmm|Granzyme M (lymphocyte met ase 1)|MET1|Met-ase|Met ase|MET 1|LMET 1|HU-Met-1|Met1 serine protease|Lymphocyte met ase 1|Met 1 serine protease|GZM M|HU Met 1|LMET1|Natural killer cell granular protease|Granzyme M|GRAM_HUMAN|Met-1 serine protease
  • GranzymeM|Gzmm|Granzyme M (lymphocyte met ase 1)|MET1|Met-ase|Met ase|MET 1|LMET 1|HU-Met-1|Met1 serine protease|Lymphocyte met ase 1|Met 1 serine protease|GZM M|HU Met 1|LMET1|Natural killer cell granular protease|Granzyme M|GRAM_HUMAN|Met-1 serine protease
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • P51124
  • P51124
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide from Human protein at AA range: 207-256
  • Synthetic peptide from Human protein at AA range: 207-256
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Antigen affinity purified
  • Antigen affinity purified
  • Storage Buffer:
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
  • PBS, 0.02% sodium azide, 50% glycerol, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together