Anti-RBP5 Rabbit Polyclonal Antibody
77704-744
New Product
- Antigen Symbol:RBP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:P82980
- Antigen Synonyms:CRBP III|Retinol binding protein 5|RET5_HUMAN|Cellular retinol-binding protein III|CRBPIII|CRBP3|Retinol-binding protein 5|RBP5|CRBP-III|Retinol binding protein|HRBPiso|cellular|OTTHUMP00000238163|Putative cellular retinol binding protein CRBP III
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the N-terminal region of Human CRBP-III
- Purification:Antigen affinity purified
- Cat. No.:77704-744
- Packaging:Vial
IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:20000
Type:
Antigen: RBP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human