Anti-TCEAL4 Rabbit Polyclonal Antibody
Catalog # 77704-638
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:TCEAL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q96EI5
- Antigen Synonyms:TCEA-like protein 4|NPD017|Transcription elongation factor S-II protein-like 4|TCEAL4|Transcription elongation factor A protein-like 4
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human TCEAL4
- Purification:Antigen affinity purified
- Cat. No.:77704-638
- Packaging:Vial
Specifications
About this item
Type:
Antigen: TCEAL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human