Anti-SH3TC2 Rabbit Polyclonal Antibody
Catalog # 77704-625
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SH3TC2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:Q8TF17
- Antigen Synonyms:PP12494|D430044G18Rik|FLJ13605|SH3TC2|CMT4C|SH3 domain and tetratricopeptide repeats-containing protein 2|MNMN|RGD1309038|S3TC2_HUMAN|KIAA1985
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human SH3TC2
- Purification:Antigen affinity purified
- Cat. No.:77704-625
- Packaging:Vial
Specifications
About this item
Type:
Antigen: SH3TC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human