Anti-ZAP70 (phospho-Y315) Rabbit Polyclonal Antibody
Catalog # 77704-569
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ZAP70 (phospho-Y315)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P43403
- Antigen Synonyms:Zeta chain associated protein kinase 70kDa isoform 1|Zeta chain TCR associated protein kinase 70kDa|Tyrosine protein kinase ZAP70|Selective T cell defect|Zeta chain associated protein kinase 70kD|FLJ17670|SRK|Tyrosine-protein kinase ZAP-70|Zeta chain associated protein kinase 70kDa|Zeta chain TCR associated protein kinase 70kD|TZK|Zeta chain of T cell receptor associated protein kinase 70|70 kDa zeta-associated protein|Truncated ZAP kinase|ZAP 70|ZAP70|70 kDa zeta associated protein|Syk-related tyrosine kinase|STD|Syk related tyrosine kinase|Zeta chain associated protein kinase 70kDa isoform 2|ZAP70_HUMAN|FLJ17679|EC 2.7.10.2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human ZAP-70 around the phosphorylation site of Y315
- Purification:Antigen affinity purified
- Cat. No.:77704-569
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: ZAP70 (phospho-Y315)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse