Anti-EZR (phospho-T566) Rabbit Polyclonal Antibody
Catalog # 77704-539
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EZR (phospho-T566)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P15311
- Antigen Synonyms:Cytovillin|Villin 2 ezrin|VIL2|Epididymis secretory protein Li 105|DKFZp762H157|EZRI_HUMAN|EZR|p81|CVIL|Ezrin|HEL S 105|FLJ26216|Villin-2|Cytovillin 2|VIL 2|MGC1584|Villin2|Villin 2 (ezrin)|Villin 2|CVL
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human Ezrin around the phosphorylation site of T566
- Purification:Antigen affinity purified
- Cat. No.:77704-539
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: EZR (phospho-T566)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat