Anti-VASP (phospho-S238) Rabbit Polyclonal Antibody
77704-521
New Product
- Antigen Symbol:VASP (phospho-S238)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P50552
- Antigen Synonyms:Vasodilator-stimulated phosphoprotein|VASP_HUMAN|VASP|Vasodilator stimulated phosphoprotein
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human VASP around the phosphorylation site of S238
- Purification:Antigen affinity purified
- Cat. No.:77704-521
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: VASP (phospho-S238)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat