Anti-STK11 (phospho-S334) Rabbit Polyclonal Antibody
Catalog # 77704-357
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:STK11 (phospho-S334)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:Q15831
- Antigen Synonyms:Serine/Threonine Kinase 11|Stk11|Serine/threonine protein kinase LKB1|Polarization related protein LKB1|PJS|Serine/threonine protein kinase 11|Renal carcinoma antigen NY-REN-19|Serine/threonine-protein kinase 11|hLKB1|LKB1|Serine/threonine protein kinase STK11|Serine/threonine-protein kinase XEEK1|Liver kinase B1|STK11_HUMAN|Serine/threonine-protein kinase LKB1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from Human LKB1 around the phosphorylation site of S334
- Purification:Antigen affinity purified
- Cat. No.:77704-357
- Packaging:Vial
Specifications
About this item
IHC:1:100-1:300, ELISA:1:10000
Type:
Antigen: STK11 (phospho-S334)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse