Anti-RREB1 Rabbit Polyclonal Antibody
Catalog # 77704-157
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:RREB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q92766
- Antigen Synonyms:Zep1|RAS responsive element binding protein 1|RREB-1|Zinc finger motif enhancer binding protein 1|Raf responsive zinc finger protein LZ321|Finger protein in nuclear bodies|Zep-1|Raf-responsive zinc finger protein LZ321|HNT|FINB|Ras-responsive element-binding protein 1|LZ321|RREB1_HUMAN|Rreb1|Zinc finger motif enhancer-binding protein 1|Hindsight homolog|RREB 1|Zep 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human RREB1
- Purification:Antigen affinity purified
- Cat. No.:77704-157
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:20000
Type:
Antigen: RREB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse